Assay Details
Gene Symbol: parp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058671.10 | NP_491072.1 | 5 - 6 | 1639 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
