Assay Details
Gene Symbol: Y54E10A.11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058717.6 | NP_491118.2 | 4 - 5 | 1430 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
