Assay Details
Gene Symbol: lin-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058805.7 | NP_491206.1 | 2 - 3 | 608 | 70 | |
GenBank mRNA | AF163019.1 | - | 1 - 2 | 173 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
