Assay Details
Gene Symbol: lin-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058805.7 | NP_491206.1 | 3 - 4 | 927 | 72 | |
GenBank mRNA | AF163019.1 | - | 2 - 3 | 492 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
