Assay Details
Gene Symbol: lin-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058805.7 | NP_491206.1 | 5 - 6 | 1786 | 126 | |
GenBank mRNA | AF163019.1 | - | 4 - 5 | 1351 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
