Assay Details
Gene Symbol: lin-59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_058805.7 | NP_491206.1 | 8 - 9 | 4305 | 69 | |
GenBank mRNA | AF163019.1 | - | 7 - 8 | 3870 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
