Assay Details
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025813.2 | NP_001020984.1 | 17 - 18 | 3725 | 59 | |
NM_001025814.4 | NP_001020985.1 | 17 - 18 | 3534 | 59 | ||
NM_001025817.4 | NP_001020988.1 | 17 - 18 | 3534 | 59 | ||
NM_001025818.3 | NP_001020989.1 | 17 - 18 | 3534 | 59 | ||
NM_001356807.3 | NP_001343717.1 | 17 | 3534 | 59 | ||
NM_001356808.3 | NP_001343716.1 | 17 | 3534 | 59 | ||
NM_001356809.3 | NP_001343714.1 | 17 | 3534 | 59 | ||
NM_001356810.3 | NP_001343718.1 | 17 | 3534 | 59 | ||
NM_001356811.3 | NP_001343715.1 | 17 | 3534 | 59 | ||
NM_001356812.3 | NP_001343617.1 | 8 - 9 | 2115 | 59 | ||
NM_001356813.3 | NP_001343719.1 | 8 - 9 | 2115 | 59 | ||
NM_001356814.3 | NP_001343711.1 | 8 - 9 | 2115 | 59 | ||
NM_001356815.3 | NP_001343712.1 | 8 - 9 | 2115 | 59 | ||
NM_001356816.3 | NP_001343713.1 | 8 - 9 | 2115 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
