Assay Details
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025813.2 | NP_001020984.1 | 23 - 24 | 9280 | 82 | |
NM_001025814.4 | NP_001020985.1 | 23 - 24 | 9089 | 82 | ||
NM_001025817.4 | NP_001020988.1 | 21 - 22 | 7169 | 82 | ||
NM_001025818.3 | NP_001020989.1 | 21 - 22 | 7169 | 82 | ||
NM_001356807.3 | NP_001343717.1 | 23 | 9089 | 82 | ||
NM_001356809.3 | NP_001343714.1 | 22 | 9023 | 82 | ||
NM_001356810.3 | NP_001343718.1 | 23 | 9089 | 82 | ||
NM_001356811.3 | NP_001343715.1 | 22 | 9023 | 82 | ||
NM_001356812.3 | NP_001343617.1 | 14 - 15 | 7670 | 82 | ||
NM_001356813.3 | NP_001343719.1 | 14 - 15 | 7670 | 82 | ||
NM_001356815.3 | NP_001343712.1 | 13 - 14 | 7604 | 82 | ||
NM_001356816.3 | NP_001343713.1 | 12 - 13 | 5750 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
