Assay Details
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025814.4 | NP_001020985.1 | 44 - 45 | 23355 | 69 | |
NM_001025818.3 | NP_001020989.1 | 42 - 43 | 21435 | 69 | ||
NM_001306524.2 | NP_001293453.1 | 10 - 11 | 3386 | 69 | ||
NM_001306525.3 | NP_001293454.1 | 10 - 11 | 3435 | 69 | ||
NM_001356807.3 | NP_001343717.1 | 43 | 21195 | 69 | ||
NM_001356808.3 | NP_001343716.1 | 44 | 21636 | 69 | ||
NM_001356809.3 | NP_001343714.1 | 43 | 23289 | 69 | ||
NM_001356812.3 | NP_001343617.1 | 35 - 36 | 21936 | 69 | ||
NM_001356813.3 | NP_001343719.1 | 34 - 35 | 19776 | 69 | ||
NM_001356814.3 | NP_001343711.1 | 35 - 36 | 20217 | 69 | ||
NM_001356815.3 | NP_001343712.1 | 34 - 35 | 21870 | 69 | ||
NM_001356816.3 | NP_001343713.1 | 33 - 34 | 20016 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
