Assay Details
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025814.4 | NP_001020985.1 | 47 - 48 | 24151 | 67 | |
NM_001025818.3 | NP_001020989.1 | 45 - 46 | 22231 | 67 | ||
NM_001306524.2 | NP_001293453.1 | 13 - 14 | 4182 | 67 | ||
NM_001306525.3 | NP_001293454.1 | 13 - 14 | 4231 | 67 | ||
NM_001356807.3 | NP_001343717.1 | 46 | 21991 | 67 | ||
NM_001356808.3 | NP_001343716.1 | 47 | 22432 | 67 | ||
NM_001356809.3 | NP_001343714.1 | 46 | 24085 | 67 | ||
NM_001356812.3 | NP_001343617.1 | 38 - 39 | 22732 | 67 | ||
NM_001356813.3 | NP_001343719.1 | 37 - 38 | 20572 | 67 | ||
NM_001356814.3 | NP_001343711.1 | 38 - 39 | 21013 | 67 | ||
NM_001356815.3 | NP_001343712.1 | 37 - 38 | 22666 | 67 | ||
NM_001356816.3 | NP_001343713.1 | 36 - 37 | 20812 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
