Assay Details
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025814.4 | NP_001020985.1 | 37 - 38 | 20707 | 73 | |
NM_001025818.3 | NP_001020989.1 | 35 - 36 | 18787 | 73 | ||
NM_001306524.2 | NP_001293453.1 | 3 - 4 | 738 | 73 | ||
NM_001306525.3 | NP_001293454.1 | 3 - 4 | 787 | 73 | ||
NM_001356807.3 | NP_001343717.1 | 36 | 18547 | 73 | ||
NM_001356808.3 | NP_001343716.1 | 37 | 18988 | 73 | ||
NM_001356809.3 | NP_001343714.1 | 36 | 20641 | 73 | ||
NM_001356812.3 | NP_001343617.1 | 28 - 29 | 19288 | 73 | ||
NM_001356813.3 | NP_001343719.1 | 27 - 28 | 17128 | 73 | ||
NM_001356814.3 | NP_001343711.1 | 28 - 29 | 17569 | 73 | ||
NM_001356815.3 | NP_001343712.1 | 27 - 28 | 19222 | 73 | ||
NM_001356816.3 | NP_001343713.1 | 26 - 27 | 17368 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
