Assay Details
Gene Symbol: unc-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025814.4 | NP_001020985.1 | 41 - 42 | 21726 | 60 | |
NM_001025818.3 | NP_001020989.1 | 39 - 40 | 19806 | 60 | ||
NM_001306524.2 | NP_001293453.1 | 7 - 8 | 1757 | 60 | ||
NM_001306525.3 | NP_001293454.1 | 7 - 8 | 1806 | 60 | ||
NM_001356807.3 | NP_001343717.1 | 40 | 19566 | 60 | ||
NM_001356808.3 | NP_001343716.1 | 41 | 20007 | 60 | ||
NM_001356809.3 | NP_001343714.1 | 40 | 21660 | 60 | ||
NM_001356812.3 | NP_001343617.1 | 32 - 33 | 20307 | 60 | ||
NM_001356813.3 | NP_001343719.1 | 31 - 32 | 18147 | 60 | ||
NM_001356814.3 | NP_001343711.1 | 32 - 33 | 18588 | 60 | ||
NM_001356815.3 | NP_001343712.1 | 31 - 32 | 20241 | 60 | ||
NM_001356816.3 | NP_001343713.1 | 30 - 31 | 18387 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
