Assay Details
Gene Symbol: acl-11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059078.5 | NP_491479.2 | 4 - 5 | 1005 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
