Assay Details
Gene Symbol: mes-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026321.4 | NP_001021492.1 | 13 - 14 | 2003 | 127 | |
NM_001374884.2 | NP_001361974.1 | 12 - 13 | 1950 | 127 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
