Assay Details
Gene Symbol: mes-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026321.4 | NP_001021492.1 | 8 - 9 | 1165 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
