Assay Details
Gene Symbol: let-363 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059148.5 | NP_491549.2 | 3 - 4 | 559 | 96 | |
NM_059151.7 | NP_491552.2 | 3 - 4 | 521 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
