Assay Details
Gene Symbol: C37C3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383347.1 | 5 - 6 | 763 | 131 |
Gene Symbol: C37C3.18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383345.1 | 5 - 6 | 756 | 131 | ||
NM_001383346.1 | 5 - 6 | 764 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
