Assay Details
Gene Symbol: C37C3.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383347.1 | 8 - 9 | 1460 | 94 |
Gene Symbol: C37C3.18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383345.1 | 8 - 9 | 1414 | 94 | ||
NM_001383346.1 | 8 - 9 | 1465 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
