Assay Details
Gene Symbol: max-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380794.1 | NP_001367137.1 | 2 - 3 | 231 | 94 | |
NM_001383435.1 | 3 - 4 | 373 | 94 | |||
GenBank mRNA | AY084080.1 | - | 2 - 3 | 231 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383435.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
