Assay Details
Gene Symbol: par-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028848.4 | NP_001024019.1 | 5 - 6 | 1337 | 101 | |
GenBank mRNA | U40858.1 | - | 5 - 6 | 1337 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
