Assay Details
Gene Symbol: mes-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171780.7 | NP_741906.1 | 1 - 2 | 142 | 76 | |
GenBank mRNA | AF200199.1 | - | 1 - 2 | 115 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
