Assay Details
Gene Symbol: mes-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171780.7 | NP_741906.1 | 6 - 7 | 1039 | 138 | |
GenBank mRNA | AF200199.1 | - | 6 - 7 | 1012 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
