Assay Details
Gene Symbol: pat-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381702.1 | NP_001367555.1 | 2 - 3 | 50 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
