Assay Details
Gene Symbol: F48E3.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373365.3 | NP_001360756.1 | 31 - 32 | 5776 | 97 | |
NM_171729.7 | NP_741841.2 | 5 - 6 | 841 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_171729.7 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
