Assay Details
Gene Symbol: B0511.12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001264526.2 | NP_001251455.1 | 25 - 26 | 6451 | 91 | |
NM_001264527.3 | NP_001251456.1 | 25 - 26 | 6541 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
