Assay Details
Gene Symbol: fkb-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_060391.4 | NP_492792.1 | 2 - 3 | 382 | 153 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
