Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 24 | 13764 | 85 | |
NM_001348621.3 | NP_001335519.1 | 25 | 14103 | 85 | ||
NM_001348622.3 | NP_001335522.1 | 20 - 21 | 14145 | 85 | ||
NM_001348623.3 | NP_001335521.1 | 21 - 22 | 14484 | 85 | ||
NM_001348624.3 | NP_001335523.1 | 10 - 11 | 9999 | 85 | ||
NM_001348625.3 | NP_001335524.1 | 8 - 9 | 9657 | 85 | ||
NM_001377639.1 | NP_001364631.1 | 26 - 27 | 14188 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
