Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 26 | 14217 | 70 | |
NM_001348621.3 | NP_001335519.1 | 27 | 14556 | 70 | ||
NM_001348622.3 | NP_001335522.1 | 22 - 23 | 14598 | 70 | ||
NM_001348623.3 | NP_001335521.1 | 23 - 24 | 14937 | 70 | ||
NM_001348624.3 | NP_001335523.1 | 12 - 13 | 10452 | 70 | ||
NM_001348625.3 | NP_001335524.1 | 10 - 11 | 10110 | 70 | ||
NM_001377639.1 | NP_001364631.1 | 28 - 29 | 14641 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
