Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 17 | 4916 | 91 | |
NM_001348621.3 | NP_001335519.1 | 18 | 5255 | 91 | ||
NM_001348622.3 | NP_001335522.1 | 13 - 14 | 5297 | 91 | ||
NM_001348623.3 | NP_001335521.1 | 14 - 15 | 5636 | 91 | ||
NM_001348624.3 | NP_001335523.1 | 3 - 4 | 1151 | 91 | ||
NM_001348625.3 | NP_001335524.1 | 1 - 2 | 809 | 91 | ||
NM_001377639.1 | NP_001364631.1 | 18 - 19 | 5307 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
