Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 18 | 6372 | 64 | |
NM_001348621.3 | NP_001335519.1 | 19 | 6711 | 64 | ||
NM_001348622.3 | NP_001335522.1 | 14 - 15 | 6753 | 64 | ||
NM_001348623.3 | NP_001335521.1 | 15 - 16 | 7092 | 64 | ||
NM_001348624.3 | NP_001335523.1 | 4 - 5 | 2607 | 64 | ||
NM_001348625.3 | NP_001335524.1 | 2 - 3 | 2265 | 64 | ||
NM_001377639.1 | NP_001364631.1 | 19 - 20 | 6763 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
