Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 19 | 7420 | 90 | |
NM_001348621.3 | NP_001335519.1 | 20 | 7759 | 90 | ||
NM_001348622.3 | NP_001335522.1 | 15 - 16 | 7801 | 90 | ||
NM_001348623.3 | NP_001335521.1 | 16 - 17 | 8140 | 90 | ||
NM_001348624.3 | NP_001335523.1 | 5 - 6 | 3655 | 90 | ||
NM_001348625.3 | NP_001335524.1 | 3 - 4 | 3313 | 90 | ||
NM_001377639.1 | NP_001364631.1 | 20 - 21 | 7811 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
