Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 20 | 7889 | 57 | |
NM_001348621.3 | NP_001335519.1 | 21 | 8228 | 57 | ||
NM_001348622.3 | NP_001335522.1 | 16 - 17 | 8270 | 57 | ||
NM_001348623.3 | NP_001335521.1 | 17 - 18 | 8609 | 57 | ||
NM_001348624.3 | NP_001335523.1 | 6 - 7 | 4124 | 57 | ||
NM_001348625.3 | NP_001335524.1 | 4 - 5 | 3782 | 57 | ||
NM_001377639.1 | NP_001364631.1 | 21 - 22 | 8280 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
