Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 21 | 11040 | 80 | |
NM_001348621.3 | NP_001335519.1 | 22 | 11379 | 80 | ||
NM_001348622.3 | NP_001335522.1 | 17 - 18 | 11421 | 80 | ||
NM_001348623.3 | NP_001335521.1 | 18 - 19 | 11760 | 80 | ||
NM_001348624.3 | NP_001335523.1 | 7 - 8 | 7275 | 80 | ||
NM_001348625.3 | NP_001335524.1 | 5 - 6 | 6933 | 80 | ||
NM_001377639.1 | NP_001364631.1 | 22 - 23 | 11431 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
