Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 22 | 11945 | 70 | |
NM_001348621.3 | NP_001335519.1 | 23 | 12284 | 70 | ||
NM_001348622.3 | NP_001335522.1 | 18 - 19 | 12326 | 70 | ||
NM_001348623.3 | NP_001335521.1 | 19 - 20 | 12665 | 70 | ||
NM_001348624.3 | NP_001335523.1 | 8 - 9 | 8180 | 70 | ||
NM_001348625.3 | NP_001335524.1 | 6 - 7 | 7838 | 70 | ||
NM_001377639.1 | NP_001364631.1 | 23 - 24 | 12336 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
