Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348620.3 | NP_001335520.1 | 23 | 13643 | 63 | |
NM_001348621.3 | NP_001335519.1 | 24 | 13982 | 63 | ||
NM_001348622.3 | NP_001335522.1 | 19 - 20 | 14024 | 63 | ||
NM_001348623.3 | NP_001335521.1 | 20 - 21 | 14363 | 63 | ||
NM_001348624.3 | NP_001335523.1 | 9 - 10 | 9878 | 63 | ||
NM_001348625.3 | NP_001335524.1 | 7 - 8 | 9536 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
