Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047256.5 | NP_001040721.1 | 13 - 14 | 3493 | 60 | |
NM_001129093.3 | NP_001122565.1 | 9 - 10 | 3874 | 60 | ||
NM_001348620.3 | NP_001335520.1 | 12 | 3154 | 60 | ||
NM_001348621.3 | NP_001335519.1 | 13 | 3493 | 60 | ||
NM_001348622.3 | NP_001335522.1 | 8 - 9 | 3535 | 60 | ||
NM_001348623.3 | NP_001335521.1 | 9 - 10 | 3874 | 60 | ||
NM_001377639.1 | NP_001364631.1 | 13 - 14 | 3545 | 60 | ||
NM_001377640.2 | NP_001364629.1 | 18 - 19 | 4303 | 60 | ||
NM_001377641.2 | NP_001364630.1 | 15 - 16 | 3949 | 60 | ||
NM_001377642.2 | NP_001364643.1 | 13 - 14 | 3400 | 60 | ||
NM_001377643.3 | NP_001364644.1 | 12 - 13 | 3213 | 60 | ||
NM_001377644.3 | NP_001364645.1 | 13 - 14 | 3493 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
