Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047256.5 | NP_001040721.1 | 5 - 6 | 505 | 66 | |
NM_001348620.3 | NP_001335520.1 | 5 | 505 | 66 | ||
NM_001348621.3 | NP_001335519.1 | 5 | 505 | 66 | ||
NM_001377639.1 | NP_001364631.1 | 5 - 6 | 557 | 66 | ||
NM_001377640.2 | NP_001364629.1 | 10 - 11 | 1315 | 66 | ||
NM_001377641.2 | NP_001364630.1 | 7 - 8 | 961 | 66 | ||
NM_001377642.2 | NP_001364643.1 | 5 - 6 | 412 | 66 | ||
NM_001377643.3 | NP_001364644.1 | 5 - 6 | 564 | 66 | ||
NM_001377644.3 | NP_001364645.1 | 5 - 6 | 505 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
