Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047256.5 | NP_001040721.1 | 8 - 9 | 879 | 79 | |
NM_001129093.3 | NP_001122565.1 | 4 - 5 | 1260 | 79 | ||
NM_001348621.3 | NP_001335519.1 | 8 | 879 | 79 | ||
NM_001348623.3 | NP_001335521.1 | 4 - 5 | 1260 | 79 | ||
NM_001377639.1 | NP_001364631.1 | 8 - 9 | 931 | 79 | ||
NM_001377640.2 | NP_001364629.1 | 13 - 14 | 1689 | 79 | ||
NM_001377641.2 | NP_001364630.1 | 10 - 11 | 1335 | 79 | ||
NM_001377642.2 | NP_001364643.1 | 8 - 9 | 786 | 79 | ||
NM_001377644.3 | NP_001364645.1 | 8 - 9 | 879 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
