Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047256.5 | NP_001040721.1 | 9 - 10 | 1214 | 62 | |
NM_001129093.3 | NP_001122565.1 | 5 - 6 | 1595 | 62 | ||
NM_001348621.3 | NP_001335519.1 | 9 | 1214 | 62 | ||
NM_001348623.3 | NP_001335521.1 | 5 - 6 | 1595 | 62 | ||
NM_001377639.1 | NP_001364631.1 | 9 - 10 | 1266 | 62 | ||
NM_001377640.2 | NP_001364629.1 | 14 - 15 | 2024 | 62 | ||
NM_001377641.2 | NP_001364630.1 | 11 - 12 | 1670 | 62 | ||
NM_001377642.2 | NP_001364643.1 | 9 - 10 | 1121 | 62 | ||
NM_001377644.3 | NP_001364645.1 | 9 - 10 | 1214 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
