Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047256.5 | NP_001040721.1 | 10 - 11 | 2743 | 95 | |
NM_001129093.3 | NP_001122565.1 | 6 - 7 | 3124 | 95 | ||
NM_001348620.3 | NP_001335520.1 | 9 | 2404 | 95 | ||
NM_001348621.3 | NP_001335519.1 | 10 | 2743 | 95 | ||
NM_001348622.3 | NP_001335522.1 | 5 - 6 | 2785 | 95 | ||
NM_001348623.3 | NP_001335521.1 | 6 - 7 | 3124 | 95 | ||
NM_001377639.1 | NP_001364631.1 | 10 - 11 | 2795 | 95 | ||
NM_001377640.2 | NP_001364629.1 | 15 - 16 | 3553 | 95 | ||
NM_001377641.2 | NP_001364630.1 | 12 - 13 | 3199 | 95 | ||
NM_001377642.2 | NP_001364643.1 | 10 - 11 | 2650 | 95 | ||
NM_001377643.3 | NP_001364644.1 | 9 - 10 | 2463 | 95 | ||
NM_001377644.3 | NP_001364645.1 | 10 - 11 | 2743 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
