Assay Details
Gene Symbol: vab-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377640.2 | NP_001364629.1 | 1 - 2 | 177 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
