Assay Details
Gene Symbol: W09G3.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_061016.8 | NP_493417.1 | 4 - 5 | 805 | 97 |
Gene Symbol: W09G3.8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383751.1 | 4 - 5 | 802 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001383751.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
