Assay Details
Gene Symbol: B0432.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313561.2 | NP_001300490.1 | 7 - 8 | 1695 | 119 | |
NM_001313562.3 | NP_001300491.1 | 6 - 7 | 1448 | 119 |
Gene Symbol: pqn-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377684.3 | NP_001364775.1 | 1 - 2 | 598 | 119 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001377684.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
