Assay Details
Gene Symbol: gcp-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029695.6 | NP_001024866.1 | 3 - 4 | 710 | 86 | |
NM_075684.7 | NP_508085.2 | 4 - 5 | 760 | 86 | ||
NM_182448.5 | NP_872248.2 | 3 - 4 | 710 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
