Assay Details
Gene Symbol: clhm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063002.5 | NP_495403.2 | 3 - 4 | 1125 | 135 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
