Assay Details
Gene Symbol: znf-598 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375249.1 | NP_001362178.1 | 12 - 13 | 2450 | 112 | |
NM_063038.5 | NP_495439.2 | 12 - 13 | 2444 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
