Assay Details
Gene Symbol: cdc-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063197.9 | NP_495598.1 | 1 - 2 | 126 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
