Assay Details
Gene Symbol: gbh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063392.3 | NP_495793.2 | 4 - 5 | 875 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
