Assay Details
Gene Symbol: mak-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027067.5 | NP_001022238.1 | 4 - 5 | 659 | 76 | |
NM_063459.5 | NP_495860.1 | 6 - 7 | 1185 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
