Assay Details
Gene Symbol: mpz-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026867.4 | NP_001022038.2 | 3 - 4 | 229 | 76 | |
NM_001026868.5 | NP_001022039.2 | 3 - 4 | 210 | 76 | ||
NM_001129129.4 | NP_001122601.1 | 1 - 2 | 51 | 76 | ||
NM_001377853.2 | NP_001364655.1 | 4 - 5 | 282 | 76 | ||
NM_001377854.2 | NP_001364657.1 | 3 - 4 | 195 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001129129.4 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
